pH Buffer Solutions, Hach
±0.02, 25 °C, 50 mL, 4.01, Red
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BUFFER SOLN RED PH 4.01 50ML Catalog #CA10199-500
Product CA10199-500 is a replacement for 10199-500 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
For pH meter standardization
Available colorless, or color-coded for easy identification.
Documents
Specifications
Specifications
- Color:Red
- Size:50 mL
- Reference temperature:25 °C
- pH:4.01
- pH accuracy:±0.02
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



