Trypsin Neutralizing Solution, PromoCell
Trypsin Neutralizing Solution, 250 mL
Catalog # 10175-290
Supplier: Promocell
: C-41120
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Trypsin is the most widely employed enzyme used for cell detachment
This solution exhibit standardized enzyme activity and contain trypsin from extra-pure lots. This solution contains 0.05% trypsin inhibitor in 0.1% BSA.
Caution: For in vitro research use only and not for diagnostic or therapeutic procedures.
Specifications
Specifications
- Description:Trypsin Neutralizing Solution
- Size:250 mL
- Cat. No.:10175-290
- Storage temperature:–20 °C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



