Order Entry
Canada
Orders LinkContactUsLinkComponent
Trypsin Neutralizing Solution, PromoCell

Trypsin Neutralizing Solution, PromoCell

Trypsin Neutralizing Solution, 125 mL
Catalog # 10175-288
Supplier:   Promocell
Supplier #:  C-41110
Trypsin Neutralizing Solution, PromoCell
Catalog # 10175-288
Supplier:  Promocell
CAS Number:  
Trypsin Neutralizing Solution, PromoCell

Trypsin Neutralizing Solution, 125 mL

CAS Number:  
Price
130,88 $
Log in for availability
Each (125ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Trypsin is the most widely employed enzyme used for cell detachment

This solution exhibit standardized enzyme activity and contain trypsin from extra-pure lots. This solution contains 0.05% trypsin inhibitor in 0.1% BSA.

 : For in vitro research use only and not for diagnostic or therapeutic procedures.

Specifications

Specifications

  • Description:
    Trypsin Neutralizing Solution
  • Size:
    125 mL
  • Cat. No.:
    10175-288
  • Storage temperature:
    –20 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.