Order Entry
Canada
Orders LinkContactUsLinkComponent
Trypsin EDTA Solution, PromoCell

Trypsin EDTA Solution, PromoCell

Trypsin/EDTA 2 (0.025%/0.01%), 250 ml
Catalog # 10172-554
Supplier:   Promocell
Supplier #:  C-41022
Trypsin EDTA Solution, PromoCell
Catalog # 10172-554
Supplier:  Promocell
CAS Number:  
Trypsin EDTA Solution, PromoCell

Trypsin/EDTA 2 (0.025%/0.01%), 250 ml

CAS Number:  
Price
$126.98
Log in for availability
Each (250ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Trypsin is the most widely employed enzyme used for cell detachment.

  • Available in different sizes
  • Store at –20 °C in the dark
  • Thawed solutions can be stored at 4 to 8 °C

PromoCell's Trypsin Solutions exhibit standardized enzyme activity and contain trypsin from extra-pure lots.

All lots have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Caution: The products are for in vitro research use only and not for diagnostic or therapeutic procedures.

Specifications

Specifications

  • Description:
    Trypsin/EDTA 2 (0.025%/0.01%)
  • Volume:
    250 ml

Specifications