Order Entry
Canada
Orders LinkContactUsLinkComponent
Dendritic Cell Medium, PromoCell

Dendritic Cell Medium, PromoCell

DC Base Medium Without Cytokines, 250 ml
Catalog # 10172-402
Supplier:   Promocell
Supplier #:  C-28053
Dendritic Cell Medium, PromoCell
Catalog # 10172-402
Supplier:  Promocell
CAS Number:  
Dendritic Cell Medium, PromoCell
Dendritic Cell Medium, PromoCell

DC Base Medium Without Cytokines, 250 ml

CAS Number:  
Price
$356.63
Log in for availability
Each (250ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The PromoCell Dendritic Cell (DC) Generation Medium has been developed for the easy and efficient generation of immature as well as fully mature myeloid Dendritic Cells from peripheral blood monocytes

The DC Generation Medium DXF is a chemically defined and xeno-free formulation for use with freshly isolated cells. It is free of all animal derived components and substances of human origin with Human Serum Albumin as the only exception. The DC Base Media do not include cytokines and must therefore be supplemented according to the user’s individual needs. The Monocyte Attachment Medium allows for the efficient adherence selection of Monocytes from freshly isolated Mononuclear Cells (MNC) while maintaining optimal cell health. Thus, the time-consuming and costly immunomagnetic purification of monocytes prior to dendritic cell generation is not necessary and can be skipped.

Recommended for Human Monocytes, freshly isolated and cryopreserved, Human Mononuclear Cells, freshly isolated

PromoCell DC Generation Medium allows for the generation of DC from cryopreserved as well as freshly isolated cells. In the latter case, the Monocyte Attachment Medium is also needed.

Ordering information: PromoCell DC Generation Media contain all growth factors and supplements. The DC Base Media come without cytokines and must therefore be adequately supplemented by the user. The Monocyte Attachment Medium does not need any further supplementation.

Specifications

Specifications

  • Description:
    DC Base Medium Without Cytokines
  • Size:
    250 ml

Specifications