Phosphate Test Strips, MQuant®
Product Details & Documents
MQuant™ test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
- Easy to handle: Just dip into sample, add reagent, wait and measure visually against colour scale
- Immediate results within 2 minutes enables fast decisions
- Portable: Can be used inside or outside the laboratory
The phosphate test is suitable for determining orthophosphate in wastewater, soil samples, fertilizers and food.
Gradation: 10 - 25 - 50 - 100 - 250 - 500 mg/l PO₄³⁻
Documents
Certificates
Search by Lot NumberEMD Millipore Simply Rapid MQuant and MColorpHast Mobile Analysis System MilliporeSigma Food and Beverage Applications
Product Literature
ghs_1.10428
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specifications
- Pack type:Plastic Box(es)
- Description:Phophate test strips
- Size:1 pc
- Range:10 - 500 mg/L PO₄³⁻
- No. of tests:100
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)