Caps for Sample Cups, Globe Scientific
Product Details & Documents
Produced from polyethylene (PE), these caps are designed specifically for use on Globe's popular line of multi-purpose sample cups
Suitable for use with the Globe Scientific Multi-Purpose Sampling Cups series.
Caps are available with either a solid or cross-cut pierceable top, and they snap securely in place on each of the sample cups.
Specifications
- Description:Flat Top Snap Caps
- Material:Polyethylene
- Thread size:GL-45
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.