Order Entry
Canada
Orders LinkContactUsLinkComponent
Sodium Nitrate

Sodium Nitrate

Sodium Nitrate, Reagent, 500 g
Catalog # 470302-634
Supplier:   Ward's Science
Supplier #:  SS0680-500GR
Sodium Nitrate
Catalog # 470302-634
Supplier:  Ward's Science
CAS Number:  
Sodium Nitrate
Sodium Nitrate

Sodium Nitrate, Reagent, 500 g

CAS Number:  
Price
$55.70
Log in for availability
Each (500g)
 
Product Details & Documents

CAS Number: 7631-99-4
Formula Weight: 84.99
Formula: NaNO3
Hazard Info: Oxidizer, Explosive, Toxic
Density (g/mL): 2.261
Freezing Point (°C): 307
Solubility: Water and Alcohol
Synonyms: Nitrate of Soda, Chile Saltpeter
Shelf Life (months): 36
Storage: Yellow

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Specifications

Specifications

  • Description:
    Sodium Nitrate
  • Size:
    500 g
  • Chemical/physical specifications:
    Crystal
  • Grade:
    Reagent

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.