Sodium Hydroxide
Sodium Hydroxide, Solution, Laboratory, 3.8 L
Supplier: Ward's Science
Supplier #: SS0600-3.8L
Jump to:
Product Details & Documents
CAS Number: 1310-73-2
Formula: NaOH
Density: 2.13 g/mL
Boiling and Freezing Point: 1390°C, 318°C
Solubility: Water and Alcohol
Synonyms: Caustic Soda
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Caution: This product is corrosive and toxic.
Documents
sds Sodium Hydroxide, 0.01 M Solution ,SS0593.pdf sds Sodium Hydroxide, 0.2 M Solution ,SS0581.pdf sds Sodium Hydroxide, 30% Solution ,SS0601.pdf sds_Sodium Hydroxide, 0.01 Molar Solution_SS0593.pdf sds_Sodium Hydroxide, 0.1 Molar (0.1N) Solution_SS0600.pdf
SDS
More
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specifications
- Description:Sodium Hydroxide, Solution
- Size:3.8 L
- Chemical/physical specifications:0.1 M
- Grade:Laboratory



