Order Entry
Canada
Orders LinkContactUsLinkComponent
Hydrochloric Acid

Hydrochloric Acid

Hydrochloric acid, concentrated, Reagent ACS, 4×2.5 L
Catalog # 470301-262
Supplier:   Ward's Science
Supplier #:  HH0171-PK/4R
Hydrochloric Acid
Catalog # 470301-262
Supplier:  Ward's Science
CAS Number:  
Hydrochloric Acid
Hydrochloric Acid

Hydrochloric acid, concentrated, Reagent ACS, 4×2.5 L

CAS Number:  
Price
$1,512.60
Log in for availability
Case of 4 (2,500,ml) Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 7647-01-0
Formula: HCl
Density: 1.16 mL
Boiling and Freezing Point: 81.11 to 85°C, -45°C
Solubility: Water
Synonyms: Muriatic Acid, Hydrogen Chloride
Shelf Life: 24 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: This product is corrosive and toxic.

Specifications

Specifications

  • Description:
    Hydrochloric acid, concentrated
  • Size:
    4×2.5 L
  • Chemical/physical specifications:
    12 M, 37%
  • Grade:
    Reagent ACS

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.