Order Entry
Canada
Orders LinkContactUsLinkComponent
Vegetable Dye

Vegetable Dye

Vegetable Dye, FD&C Blue #2, Laboratory, 50 mL
Catalog # 470300-998
Supplier:   Ward's Science
Supplier #:  FF0410-50ML
Vegetable Dye
Catalog # 470300-998
Supplier:  Ward's Science
CAS Number:  
Vegetable Dye
Vegetable Dye

Vegetable Dye, FD&C Blue #2, Laboratory, 50 mL

CAS Number:  
Price
$19.75
Log in for availability
Each (50ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Solubility: Water
Shelf Life (months): 12
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Specifications

Specifications

  • Description:
    Vegetable Dye, FD&C Blue #2
  • Size:
    50 mL
  • Chemical/physical specifications:
    Blue #2
  • Grade:
    Laboratory

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.