Order Entry
Canada
Orders LinkContactUsLinkComponent
Silver Nitrate

Silver Nitrate

Silver Nitrate, QuickPrep, Laboratory, Makes 1 L
Catalog # 470302-378
Supplier:   Ward's Science
Supplier #:  SS0160-CAPS
Silver Nitrate
Catalog # 470302-378
Supplier:  Ward's Science
CAS Number:  
Silver Nitrate
Silver Nitrate

Silver Nitrate, QuickPrep, Laboratory, Makes 1 L

CAS Number:  
Price
$525.00
Log in for availability
Each (50ml)
Minimum order quantity is 1
Product Details & Documents

CAS Number: 7761-88-8
Formula: AgNO3
Density: 4.35 g/mL
Freezing Point: 212 °C
Solubility: Water, Hot Alcohol
Shelf Life: 24 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: This product is a corrosive, toxic irritant.

Specifications

Specifications

  • Description:
    Silver Nitrate, QuickPrep
  • Size:
    Makes 1 L
  • Chemical/physical specifications:
    0.1 M
  • Grade:
    Laboratory

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.