Order Entry
Canada
Orders LinkContactUsLinkComponent
Potassium Nitrate

Potassium Nitrate

Potassium Nitrate, QuickPrep, Laboratory, Makes 500 mL
Catalog # 470302-192
Supplier:   Ward's Science
Supplier #:  PP0662-CONC
Potassium Nitrate
Catalog # 470302-192
Supplier:  Ward's Science
CAS Number:  
Potassium Nitrate
Potassium Nitrate

Potassium Nitrate, QuickPrep, Laboratory, Makes 500 mL

CAS Number:  
Price
$16.75
Log in for availability
Each (50ml)
Minimum order quantity is 1
Product Details & Documents

CAS Number: 7757-79-1
Formula: KNO3
Density: 2.1 g/mL
Boiling and Freezing Point: 400°C, 333°C
Solubility: Water, Glycerin, and Alcohol
Synonyms: Potash Nitrate, Saltpeter
Shelf Life: 36 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: This product is flammable oxidizer and irritant.

Specifications

Specifications

  • Description:
    Potassium Nitrate, QuickPrep
  • Size:
    Makes 500 mL
  • Chemical/physical specifications:
    0.1 M
  • Grade:
    Laboratory

Specifications