Order Entry
Canada
Orders LinkContactUsLinkComponent
Potassium Iodate

Potassium Iodate

Potassium Iodate, Solution, Laboratory, 500 mL
Catalog # 470302-150
Supplier:   Ward's Science
Supplier #:  PP0603-500ML
Potassium Iodate
Catalog # 470302-150
Supplier:  Ward's Science
CAS Number:  
Potassium Iodate
Potassium Iodate

Potassium Iodate, Solution, Laboratory, 500 mL

CAS Number:  
Price
$19.50
Log in for availability
Each (500ml)
Minimum order quantity is 1
Product Details & Documents

CAS Number: 7758-05-6
Formula Weight: 214.00
Formula: KIO3
Hazard Info: Oxidizer, Toxic
Density (g/mL): 3.98
Solubility: Water and Dilute Sulfuric Acid
Synonyms: Iodic Acid, Potassium Salt
Shelf Life (months): 36
Storage: Yellow

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Specifications

Specifications

  • Description:
    Potassium Iodate, Solution
  • Size:
    500 mL
  • Chemical/physical specifications:
    0.1 M
  • Grade:
    Laboratory

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.