Order Entry
Canada
Orders LinkContactUsLinkComponent
Potassium Hydroxide

Potassium Hydroxide

Potassium Hydroxide, Laboratory, 500 g
Catalog # 470302-140
Supplier:   Ward's Science
Supplier #:  PP0570-500G
Potassium Hydroxide
Catalog # 470302-140
Supplier:  Ward's Science
CAS Number:  
Potassium Hydroxide
Potassium Hydroxide

Potassium Hydroxide, Laboratory, 500 g

CAS Number:  
Price
$59.00
Log in for availability
Each (500g)
Minimum order quantity is 1
Product Details & Documents

CAS Number: 1310-58-3
Formula Weight: 56.11
Formula: KOH
Hazard Info: Corrosive, Irritant, Toxic
Density (g/mL): 2.044
Boiling Point (°C): 1320
Freezing Point (°C): 361
Solubility: Water, Alcohol, and Glycerin
Synonyms: Caustic Potash, Potassium Hydrate
Shelf Life (months): 36
Storage: White

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Specifications

Specifications

  • Description:
    Potassium Hydroxide
  • Size:
    500 g
  • Chemical/physical specifications:
    Pellet
  • Grade:
    Laboratory

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.