Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Potassium Bromide

Potassium Bromide, Reagent, 100 g
Catalog # 470302-064
Supplier:   Ward's Science
 :  PP0400-100GR
Potassium Bromide
Catalog # 470302-064
Supplier:  Ward's Science
CAS Number:  
Potassium Bromide
Potassium Bromide

Potassium Bromide, Reagent, 100 g

CAS Number:  
Price
$32.55
Log in for availability
Each (100g)
 
Product Details & Documents

CAS Number: 7758-02-3
Formula Weight: 119.01
Formula: KBr
Density (g/mL): 2.749
Boiling Point (°C): 1435
Freezing Point (°C): 760
Solubility: Water and Alcohol
Shelf Life (months): 12
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Specifications

Specifications

  • Description:
    Potassium Bromide
  • Size:
    100 g
  • Chemical/physical specifications:
    Crystal
  • Grade:
    Reagent

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.