Potassium Bromide
Potassium Bromide, Laboratory, 500 g
Supplier: Ward's Science
Supplier #: PP0400-500G
Jump to:
Product Details & Documents
CAS Number: 7758-02-3
Formula Weight: 119.01
Formula: KBr
Density (g/mL): 2.749
Boiling Point (°C): 1435
Freezing Point (°C): 760
Solubility: Water and Alcohol
Shelf Life (months): 12
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Specifications
Specifications
- Description:Potassium Bromide
- Size:500 g
- Chemical/physical specifications:Crystal
- Grade:Laboratory
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



