Order Entry
Canada
Orders LinkContactUsLinkComponent
Potassium Bromide

Potassium Bromide

Potassium Bromide, QuickPrep, Laboratory, Makes 500 mL
Catalog # 470302-058
Supplier:   Ward's Science
Supplier #:  PP0409-CONC
Potassium Bromide
Catalog # 470302-058
Supplier:  Ward's Science
CAS Number:  
Potassium Bromide
Potassium Bromide

Potassium Bromide, QuickPrep, Laboratory, Makes 500 mL

CAS Number:  
Price
$13.05
Log in for availability
Each (50ml)
Minimum order quantity is 1
Product Details & Documents

CAS Number: 7758-02-3
Formula Weight: 119.01
Formula: KBr
Density (g/mL): 2.749
Boiling Point (°C): 1435
Freezing Point (°C): 760
Solubility: Water and Alcohol
Shelf Life (months): 12
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Specifications

Specifications

  • Description:
    Potassium Bromide, QuickPrep
  • Size:
    Makes 500 mL
  • Chemical/physical specifications:
    0.1 M
  • Grade:
    Laboratory

Specifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.