Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Sodium Silicate

Sodium Silicate, Water Glass Solution, Laboratory, 500 mL
  470302-680
 :  SS0820-500ML
Sodium Silicate
  470302-680
 :  
Sodium Silicate
Sodium Silicate

Sodium Silicate, Water Glass Solution, Laboratory, 500 mL

 :  
 
$15.40
   
Each (500ml)
 
   
 

CAS Number: 10213-79-3
Formula Weight: 212.03
Formula: Na2SiO3·5H2O
Density (g/mL): 1.81
Freezing Point (°C): 72.2
Synonyms: Disodium Trioxosilicate Pentahydrate
Shelf Life (months): 36
Storage: White

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

 : This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

 

 

  • Description:
    Sodium Silicate, Water Glass Solution
  • Size:
    500 mL
  • Chemical/physical specifications:
    37 - 40% aqueous
  • Grade:
    Laboratory

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.