Order Entry
Canada
Orders LinkContactUsLinkComponent
Propidium Iodide

Propidium Iodide

500 Tests
Catalog # 10140-984
Supplier:   Tonbo Biosciences
Supplier #:  13-6990-T500
Propidium Iodide
Catalog # 10140-984
Supplier:  Tonbo Biosciences
CAS Number:  25535-16-4
Propidium Iodide
Propidium Iodide

500 Tests

CAS Number:  25535-16-4
Price
$109.57
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Propidium Iodide (PI) is a membrane-impermeant DNA binding dye that cannot penetrate viable cells. PI rapidly enters cells with compromised membranes and intercalates between base pairs allowing exclusion of non-viable cells from analysis of flow cytometry data. PI has a broad emission spectrum from 535-617 nm and can be detected in either FL2 or FL3 detectors. When used with Annexin V FITC, it is recommended to analyze PI in FL2.

  • Formulation: 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Synonyms:  3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
  • CAS Number:25535-16-4
  • Formula:C₂₇H₃₄I₂N₄
  • MDL Number:MFCD00011921
  • Molecular Weight:668.4 g/mol
Specifications

Specifications

  • Size:
    500 Tests

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.