Order Entry
Canada
Orders LinkContactUsLinkComponent
Polypropylene Bouffant Caps, High-Tech Conversions

Polypropylene Bouffant Caps, High-Tech Conversions

Blue, 19"
Catalog # 10135-736
Small Business Enterprise
Supplier #:  GAH-BP-19-BL
Polypropylene Bouffant Caps, High-Tech Conversions
Catalog # 10135-736
Small Business Enterprise
Polypropylene Bouffant Caps, High-Tech Conversions
Polypropylene Bouffant Caps, High-Tech Conversions

Blue, 19"

Price
$22.61
Log in for availability
Pack of 100 Case of 1000
Product Details & Documents

Polypropylene Bouffant Caps are highly breathable and low-linting. They provide a cost effective solution for general applications.

  • 100% Latex Free
  • Lint Free
  • Available in Blue and White
  • 100% Spunbound Polypropylene Construction
  • Soft and Comfortable
  • Equipped with Elastic Bands for Secure Fit

Bouffant caps are ideal for medical or light industrial applications, as well as applications in the food industry. Caps are designed for use in laboratories and manufacturing environments.

Ordering information: All Bouffant Caps are packaged 100 per bag, 10 bags per case.

Specifications
  • Ø:
    19"
  • Color:
    Blue
  • Cat. No.:
    10135-736
  • Fabric:
    Polypropylene
  • Weight:
    10 g/m²
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.