Order Entry
Canada
Orders LinkContactUsLinkComponent
Treston Lab Carts

Treston Lab Carts

2 shelf lab cart, 330 lbs.
Catalog # 10129-034
Supplier:   Treston
Supplier #:  TRTA4082
Treston Lab Carts
Catalog # 10129-034
Supplier:  Treston
Treston Lab Carts
Treston Lab Carts

2 shelf lab cart, 330 lbs.

Price
$1,499.52
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

TRTA lab carts are ideal for storing medical devices, instruments and supplies, and moving them to where they are needed.

  • Versatile and multi-purpose
  • Physically ergonomic for effortless cart movement

The height adjustment range of the shelves is 7.87 to 27.56".

Ordering information: Installation is required. Assembly videos available.

Specifications
  • Description:
    2 shelf lab cart
  • Load capacity:
    330 lbs.
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Extra Shelf for 4 shelf and 2 shelf trolleys
    Extra Shelf for 4 shelf and 2 shelf trolleys
    10129-048
    Log in for availability
    Each - $449.10

Product Information

  • Catalog #
    Availability
    Price
  • End mesh panels (2)
    End mesh panels (2)
    10129-050
    Log in for availability
    Each - $238.47

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.