Order Entry
Canada
Orders LinkContactUsLinkComponent
ACK Lysing Buffer, Quality Biological

ACK Lysing Buffer, Quality Biological

ACK lysing buffer, 4× 100 ml
Catalog # 10128-806
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Supplier #:  118-156-721
ACK Lysing Buffer, Quality Biological
Catalog # 10128-806
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
ACK Lysing Buffer, Quality Biological
ACK Lysing Buffer, Quality Biological

ACK lysing buffer, 4× 100 ml

CAS Number:  
Price
$454.85
Log in for availability
Pack of 4 (400ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Ammonium-Chloride-Potassium (ACK) Lysing Buffer is used for the lysis of red blood cells in samples containing white blood cells, such as EDTA-treated whole blood, buffy coats, and bone marrow.

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.

Caution: Research use only.

Specifications

Specifications

  • Description:
    ACK lysing buffer
  • Volume:
    4× 100 ml
  • Cat. No.:
    10128-806
  • pH:
    7.4 ±0.2
  • Shelf life:
    24 months
  • Sterility:
    Sterile filtered
  • Osmolality:
    290 ±5% mOsm/Kg H₂O
  • Shipping temperature:
    Ambient

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.