Order Entry
Canada
Orders LinkContactUsLinkComponent
 

ACK Lysing Buffer, Quality Biological

ACK lysing buffer, 10× 500 ml
Catalog # 10128-804
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  118-156-101CS
ACK Lysing Buffer, Quality Biological
Catalog # 10128-804
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
ACK Lysing Buffer, Quality Biological
ACK Lysing Buffer, Quality Biological

ACK lysing buffer, 10× 500 ml

CAS Number:  
Price
$1,262.95
Log in for availability
Case of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Ammonium-Chloride-Potassium (ACK) Lysing Buffer is used for the lysis of red blood cells in samples containing white blood cells, such as EDTA-treated whole blood, buffy coats, and bone marrow.

 : Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.

 : Research use only.

Specifications

Specifications

  • Description:
    ACK lysing buffer
  • Volume:
    10× 500 ml
  • Cat. No.:
    10128-804
  • pH:
    7.4 ±0.2
  • Shelf life:
    24 months
  • Sterility:
    Sterile filtered
  • Osmolality:
    290 ±5% mOsm/Kg H₂O
  • Shipping temperature:
    Ambient

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.