Order Entry
Canada
Orders LinkContactUsLinkComponent
TBS 10X Buffer Solution, pH 7.4, Quality Biological

TBS 10X Buffer Solution, pH 7.4, Quality Biological

TBS 10X pH 7.4, 10 L, 7.4 ±0.1
Catalog # 10128-702
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Supplier #:  351-086-151
TBS 10X Buffer Solution, pH 7.4, Quality Biological
Catalog # 10128-702
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
TBS 10X Buffer Solution, pH 7.4, Quality Biological
TBS 10X Buffer Solution, pH 7.4, Quality Biological

TBS 10X pH 7.4, 10 L, 7.4 ±0.1

CAS Number:  
Price
$843.05
Log in for availability
Each (10l)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

TBS 10X pH 7.4 is used as a washing buffer for cellular and molecular biology techniques.

  • DNase, RNase, and Protease tested

TBS 10X pH 7.4 will be useful as a washing buffer for electrodes, microtiter plates, and alkaline phophatase or peroxidase conjugates. TBS 10X pH 7.4 can also be used in detergent preparation and Western blot or ELISA applications.

Contenst: Tris base, Tris HCl, sodium chloride and Cell Culture Grade Water.

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.

Caution: Research use only.

Specifications

Specifications

  • Description:
    TBS 10X pH 7.4
  • Size:
    10 L
  • pH:
    7.4 ±0.1
  • Cat. No.:
    10128-702
  • Test description:
    DNAse, RNAse and protease tested
  • Shelf life:
    36 months
  • Sterility:
    Sterile filtered
  • Shipping temperature:
    Ambient

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.