TBS 10X Buffer Solution, pH 7.4, Quality Biological
TBS 10X pH 7.4, 10× 500 ml, 7.4 ±0.1
Catalog # 10128-548
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Supplier: QUALITY BIOLOGICAL, INC.
Supplier #: 351-086-101CS
TBS 10X Buffer Solution, pH 7.4, Quality Biological
Catalog # 10128-548
Supplier: QUALITY BIOLOGICAL, INC.
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
TBS 10X pH 7.4 is used as a washing buffer for cellular and molecular biology techniques.
- DNase, RNase, and Protease tested
TBS 10X pH 7.4 will be useful as a washing buffer for electrodes, microtiter plates, and alkaline phophatase or peroxidase conjugates. TBS 10X pH 7.4 can also be used in detergent preparation and Western blot or ELISA applications.
Contenst: Tris base, Tris HCl, sodium chloride and Cell Culture Grade Water.
: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
: Research use only.
Specifications
Specifications
- Description:TBS 10X pH 7.4
- Size:10× 500 ml
- pH:7.4 ±0.1
- Cat. No.:10128-548
- Test description:DNAse, RNAse and protease tested
- Shelf life:36 months
- Sterility:Sterile filtered
- Shipping temperature:Ambient



