Sodium Nitrate
Sodium Nitrate, Solution, Laboratory, 500 mL
Supplier: Ward's Science
Supplier #: SS0691-500ML
Jump to:
Product Details & Documents
CAS Number: 7631-99-4
Formula Weight: 84.99
Formula: NaNO3
Hazard Info: Oxidizer, Explosive, Toxic
Density (g/mL): 2.261
Freezing Point (°C): 307
Solubility: Water and Alcohol
Synonyms: Nitrate of Soda, Chile Saltpeter
Shelf Life (months): 36
Storage: Yellow
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Specifications
Specifications
- Description:Sodium Nitrate, Solution
- Size:500 mL
- Chemical/physical specifications:0.1 M
- Grade:Laboratory



