Order Entry
Canada
Orders LinkContactUsLinkComponent
Gram's iodine solution stabilised

Gram's iodine solution stabilised

1 gallon
Catalog # CA89407-622
Supplier #:  I128
Gram's iodine solution stabilised
Catalog # CA89407-622
Supplier:  
CAS Number:  12298-68-9
Gram's iodine solution stabilised
Gram's iodine solution stabilised

1 gallon

CAS Number:  12298-68-9
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
VWR GRAM STAIN STABILIZED IODINE GALLON Catalog #10143-180

Product 10143-180 is similar to CA89407-622 but manufacturer and specifications may vary.

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

A mordant used in the gram stain procedure. A stabilized, thicker iodine solution. Use with the intermediate decolorizer (Cat. no. D008) or fast decolorizer (Cat. no. D008F).

  • Ready to use
  • No ampules
  • A longer shelf life
Synonyms:  Lugol's iodine, Iodine and potassium iodide, Potassium triiodide
  • CAS Number:12298-68-9
  • Formula:I₃K
  • MDL Number:MFCD00134128
  • Molecular Weight:419.812 g/mol
Specifications

Specifications

  • Size:
    1 gallon

Specifications