Gram's iodine solution stabilised
8 oz. (250 ml)
Catalog # CA89407-620
Supplier: HARDY DIAGNOSTICS CA
Supplier #: I008
Gram's iodine solution stabilised
Catalog # CA89407-620
Supplier: HARDY DIAGNOSTICS CA
CAS Number: 12298-68-9
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
A mordant used in the gram stain procedure. A stabilized, thicker iodine solution. Use with the intermediate decolorizer (Cat. no. D008) or fast decolorizer (Cat. no. D008F).
- Ready to use
- No ampules
- A longer shelf life
Synonyms: Lugol's iodine, Iodine and potassium iodide, Potassium triiodide
- CAS Number:12298-68-9
- Formula:I₃K
- MDL Number:MFCD00134128
- Molecular Weight:419.812 g/mol
Documents
Gram Stain Kits and Reagents
Additional Product Details
Specifications
Specifications
- Size:8 oz. (250 ml)



