Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Fluid A, USP

Fluid A, USP, —, 200 ml, Serum bottle, 10
Catalog # CA89407-529
Fluid A, USP
Catalog # CA89407-529
Supplier:  
CAS Number:  
Fluid A, USP
Fluid A, USP

Fluid A, USP, —, 200 ml, Serum bottle, 10

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
FLUID A (SEPTUM/CAP)300ML PK10 Catalog #CA90004-128

 

Product Details & Documents

A rinsing and diluting fluid for use in USP testing protocols.

A rinsing and diluting fluid for use in USP testing protocols. Contains peptone 0.1%.

Hardy Diagnostics' sterility media and rinse solutions are manufactured in an ISO 13485 environmentally controlled production center. Each lot is certified for pH, sterility and growth promotion using ATCC® strains specified by the USP. Our approach to manufacturing ensures all sterility media and rinse fluids provide the highest level of clarity for improved reading accuracy, thus significantly reducing the risk of incorrect interpretation and false results.

Ordering information: Order by the pack of 10

Specifications

Specifications

  • Pack type:
    Serum bottle
  • Description:
    Fluid A, USP
  • Size:
  • Minimum Order Quantity:
    10
  • Volume:
    200 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.