Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Butterfield's Phosphate Buffer, Hardy Diagnostics

pH 7.2, 20×9 ml, PP tube, 13 ml
Catalog # CA89407-412
 :  K119
Butterfield's Phosphate Buffer, Hardy Diagnostics
Catalog # CA89407-412
CAS Number:  
Butterfield's Phosphate Buffer, Hardy Diagnostics
Butterfield's Phosphate Buffer, Hardy Diagnostics

pH 7.2, 20×9 ml, PP tube, 13 ml

CAS Number:  
Price
$370.00
Log in for availability
Pack of 20
 
Specifications

Specifications

  • Pack type:
    PP tube, 13 ml
  • Description:
    pH 7.2
  • Size:
    20×9 ml
  • Cat. No.:
    CA89407-412
  • Storage temperature:
    –2-30°C

Specifications