CRITERION™ Phosphate Buffer, pH 7.2
500 g, Bottle, wide mouth
Supplier: HARDY DIAGNOSTICS CA
Supplier #: C8331
Jump to:
Product Details & Documents
CRITERION™ Agar, bacteriological grade is used as a solidifying agent in the preparation of solid and semi-solid culture media.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- Increased solubility - dissolves quickly
- Custom and bulk products available - to your specifications
- BSE-free
Specifications
Specifications
- Pack type:Bottle, wide mouth
- Size:500 g
- Application:Breweries, wineries and distilleries, cannabis QA testing, clinical, cosmetics, environmental, food safety, compounding pharmacy, pharmaceutical manufacturing
- Cat. No.:CA89406-750
- Storage temperature:2...30 °C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



