Staphylococcus Select Agar, CRITERION™, Hardy Diagnostics
10 kg, Bucket
Catalog # CA89406-084
Supplier: HARDY DIAGNOSTICS CA
Supplier #: C7003
Staphylococcus Select Agar, CRITERION™, Hardy Diagnostics
Catalog # CA89406-084
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ Staphylococcus Selective Agar is a selective medium used for the isolation and differentiation of pathogenic staphylococci
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Documents
Hardy Diagnostics Criterion Dehydrated Culture Media
Product Literature
Specifications
Specifications
- Pack type:Bucket
- Size:10 kg
- Cat. No.:CA89406-084
- Storage temperature:–2-30°C



