Order Entry
Canada
Orders LinkContactUsLinkComponent
Selective Strep Agar Base, CRITERION™, Hardy Diagnostics

Selective Strep Agar Base, CRITERION™, Hardy Diagnostics

2 kg, Bucket
Catalog # CA89406-018
Supplier #:  C6892
Selective Strep Agar Base, CRITERION™, Hardy Diagnostics
Catalog # CA89406-018
CAS Number:  
Selective Strep Agar Base, CRITERION™, Hardy Diagnostics
Selective Strep Agar Base, CRITERION™, Hardy Diagnostics

2 kg, Bucket

CAS Number:  
Price
$1,220.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

CRITERION™ Selective Strep Agar Base with added sheep blood, this medium is recommended for the selective isolation and differentiation of pathogenic streptococci

  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications

Dehydratedculture media with engineered container.

Certifications: BSE Free

Specifications

Specifications

  • Pack type:
    Bucket
  • Size:
    2 kg
  • Cat. No.:
    CA89406-018
  • Storage temperature:
    –2-30°C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.