Peptic Digest Agar Base, CRITERION™, Hardy Diagnostics
10 kg, Bucket
Catalog # CA89405-864
Supplier: HARDY DIAGNOSTICS CA
: C6563
Peptic Digest Agar Base, CRITERION™, Hardy Diagnostics
Catalog # CA89405-864
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ Peptic Digest Agar Base, an enrichment medium recommended for the selective isolation of Haemophilus spp
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
: BSE Free
Specifications
Specifications
- Pack type:Bucket
- Size:10 kg
- Cat. No.:CA89405-864
- Storage temperature:–2-30°C



