Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics
10 kg, Bucket
Catalog # CA89405-368
Supplier: HARDY DIAGNOSTICS CA
Supplier #: C5573
Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics
Catalog # CA89405-368
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ Dermatophyte Test Medium is primarily for use in the isolation and detection of pathogenic dermatophytic fungi
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Documents
Certificates
Search by Lot NumberHardy Diagnostics Criterion Dehydrated Culture Media
Product Literature
Specifications
Specifications
- Pack type:Bucket
- Size:10 kg
- Cat. No.:CA89405-368
- Storage temperature:–2-30°C



