Order Entry
Canada
Orders LinkContactUsLinkComponent
Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics

Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics

500 g, Bottle, wide mouth
Catalog # CA89405-364
Supplier #:  C5571
Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics
Catalog # CA89405-364
CAS Number:  
Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics
Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics

500 g, Bottle, wide mouth

CAS Number:  
Price
$934.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

CRITERION™ Dermatophyte Test Medium is primarily for use in the isolation and detection of pathogenic dermatophytic fungi

  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications

Dehydratedculture media with engineered container.

Certifications: BSE Free

Specifications

Specifications

  • Pack type:
    Bottle, wide mouth
  • Size:
    500 g
  • Cat. No.:
    CA89405-364
  • Storage temperature:
    –2-30°C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.