Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics
122.4 g, 2 L, Zip-pouch to make 2 L, Mylar®
Supplier: HARDY DIAGNOSTICS CA
Supplier #: C5570
Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics
Catalog # CA89405-362
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ Dermatophyte Test Medium is primarily for use in the isolation and detection of pathogenic dermatophytic fungi
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Documents
Certificates
Search by Lot NumberHardy Diagnostics Criterion Dehydrated Culture Media
Product Literature
Specifications
Specifications
- Pack type:Zip-pouch to make 2 L, Mylar®
- Size:122.4 g
- Final volume:2 L
- Cat. No.:CA89405-362
- Storage temperature:–2-30°C



