Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Lead

Lead, 8×10×0.03125", Laboratory, 503 g
  470301-546
 :  LL0082-503G
Lead
  470301-546
 :  
Lead
Lead

Lead, 8×10×0.03125", Laboratory, 503 g

 :  
 
$56.70
   
Each (503g)
 
   
 

CAS Number: 7439-92-1
Formula Weight: 207.19
Formula: Pb
Hazard Info: Toxic
Density (g/mL): 11.34
Boiling Point (°C): 1753
Freezing Point (°C): 327
Solubility: Dilute Nitric Acid
Shelf Life (months): 36
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

 : This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

 

 

  • Description:
    Lead, 8×10×0.03125"
  • Size:
    503 g
  • Chemical/physical specifications:
    Sheet
  • Grade:
    Laboratory

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.