Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Iron

Iron, Laboratory, 2.5 kg
Catalog # 470301-444
Supplier:   Ward's Science
 :  IX0215-2.5KG
Iron
Catalog # 470301-444
Supplier:  Ward's Science
CAS Number:  
Iron
Iron

Iron, Laboratory, 2.5 kg

CAS Number:  
Price
$250.45
Log in for availability
Each (2,500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 65997-19-5
Formula: Fe
Density: 7.86 g/mL
Freezing Point: 1535 °C
Solubility: Sulfuric Acid and Hydrochloric Acid
Synonyms: Iron Aggregate
Shelf Life: 36 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

 : This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Specifications

Specifications

  • Description:
    Iron
  • Size:
    2.5 kg
  • Chemical/physical specifications:
    Powder
  • Grade:
    Laboratory

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.