Order Entry
Canada
Orders LinkContactUsLinkComponent
Certipur® Buffer Solution Sachet Sets, MilliporeSigma

Certipur® Buffer Solution Sachet Sets, MilliporeSigma

Certipur® buffer solution, set 1, Sachet, 30 x 30 mL, 4.01/7.00/9.00
Catalog # EM1.99005.0001
Certipur® Buffer Solution Sachet Sets, MilliporeSigma
Catalog # EM1.99005.0001
Supplier:  
CAS Number:  
Certipur® Buffer Solution Sachet Sets, MilliporeSigma
Certipur® Buffer Solution Sachet Sets, MilliporeSigma

Certipur® buffer solution, set 1, Sachet, 30 x 30 mL, 4.01/7.00/9.00

CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

30 ml buffers sachets - ideal for the calibration of pH meters.

  • Practical 30 mL sachets – one use per sachet – no risk of contamination
  • Solutions are always fresh and ready to use
  • Batch traceable to NIST
  • Reference temperature: 25°C
  • Precision: ±0.015 pH units max.
Specifications

Specifications

  • Pack type:
    Sachet
  • Description:
    Certipur® buffer solution, set 1
  • Size:
    30 x 30 mL
  • pH:
    4.01/7.00/9.00

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.