$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
CRUCIBLE PORCELAIN HIGH FORM 100ML CS24
Catalog # CA10123-936
Supplier:
Anachemia Mining
Expand image
CRUCIBLE PORCELAIN HIGH FORM 100ML CS24
Catalog # CA10123-936
Supplier:
Anachemia Mining
Supplier Number:
016-153-14
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma