Order Entry
Canada
Orders LinkContactUsLinkComponent
Hayflick Medium Agar

Hayflick Medium Agar

Hayflick medium agar, 10/pack, 11 ml, Plate, small, 15×60 mm, 10
Catalog # 10124-546
Supplier:   Hardy Diagnostics
Supplier #:  G385
Hayflick Medium Agar
Catalog # 10124-546
Supplier:  Hardy Diagnostics
CAS Number:  
Hayflick Medium Agar
Hayflick Medium Agar

Hayflick medium agar, 10/pack, 11 ml, Plate, small, 15×60 mm, 10

CAS Number:  
Price
$143.00
Log in for availability
Each
Minimum order quantity is 10
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Recommended for use in the cultivation and isolation of Mycoplasma species.

Ordering information: Minimum order of 10 packs

Documents

Specifications

Specifications

  • Description:
    Hayflick medium agar
  • Dimensions:
    15×60 mm
  • Size:
    10/pack
  • Minimum Order Quantity:
    10
  • Format:
    Plate, small
  • Application:
    Clinical
  • Volume:
    11 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.