Order Entry
Canada
Orders LinkContactUsLinkComponent
Chlorophenol Red

Chlorophenol Red

Chlorophenol Red Solution, Aqueous, 0.4%, 100 mL Bottle
Catalog # 470300-706
Supplier:   Ward's Science
Supplier #:  CC0315-100ML
Chlorophenol Red
Catalog # 470300-706
Supplier:  Ward's Science
CAS Number:  
Chlorophenol Red
Chlorophenol Red

Chlorophenol Red Solution, Aqueous, 0.4%, 100 mL Bottle

CAS Number:  
Price
$55.00
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 4430-20-0
Formula: C19H12Cl2O5S
Formula Weight: 423.27
Shelf Life (months): 36
Solubility: Water
Storage: Green

Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Specifications

Specifications

  • Description:
    Chlorophenol Red Solution, Aqueous, 0.4%, 100 mL Bottle

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.