Orders LinkContactUsLinkComponent
MicroGenesis™ Wipes, Contec®
MicroGenesis™ Wipes, Contec®
MicroGenesis™ Wipes, Bulk, Microfiber, 25.4×30.5 cm (12×10"), 100 Wipes/Bag, 8 Bags/Case
Catalog # CA89412-350
Supplier:   Contec
Supplier #:  LWMM0100
MicroGenesis™ Wipes, Contec®
Catalog # CA89412-350
Supplier:  Contec
MicroGenesis™ Wipes, Contec®
MicroGenesis™ Wipes, Contec®
MicroGenesis™ Wipes, Bulk, Microfiber, 25.4×30.5 cm (12×10"), 100 Wipes/Bag, 8 Bags/Case
Price
$787.00
Log in for availability
Case of 800
Product Details & Documents
MicroGenesis™ wipes deliver the maximum amount of microfiber to pick up and retain particles, dust, and other contaminants in cleanrooms and other critical applications.
  • Recommended for ISO Class 6-8 environments
  • Microfiber is only applied to the wiping surface creating a cost effective single use microfiber wipe
  • Microfiber fabric easily picks up and retains particles
  • Highly sorbent and suitable for use with a wide range of disinfectants
For dry applications where particle pickup and retention is critical MicroGenesis™ Wipes deliver unprecedented performance. The wipe is ideal for picking up the powder residue generated in tablet and fine powder pharmaceutical manufacturing or grease removal before manufacturing batches.
This particle pickup makes the wipe useful for surface preparation prior to painting, laminating, or other finishing applications. In addition, unique processing gives the MicroGenesis™ Wipe excellent sorbency characteristics, making it ideal for multiple applications and industries. It is safe for use with all disinfectants including chlorine and quaternary ammonium-based products.
MicroGenesis™ wipes are not recommended for applications with rough or perforated surfaces.
Specifications
  • Description:
    MicroGenesis™ Wipes, Bulk
  • Material:
    Microfiber
  • Packaging:
    100 Wipes/Bag, 8 Bags/Case
  • W×L:
    25.4×30.5 cm (12×10")