Orders LinkContactUsLinkComponent
TBD-1™ Decalcifier, Epredia
TBD-1™ Decalcifier, Epredia
Epredia™ TBD-1™ decalcifier, 2 x 1 l
Catalog # CA89369-400
Supplier:   Thermo Scientific
Supplier #:  6764001
TBD-1™ Decalcifier, Epredia
Catalog # CA89369-400
Supplier:  Thermo Scientific
CAS Number:  
TBD-1™ Decalcifier, Epredia
TBD-1™ Decalcifier, Epredia
Epredia™ TBD-1™ decalcifier, 2 x 1 l
CAS Number:  
Price
$350.06
Log in for availability
Case of 2 (0.5.l)
Product Details & Documents
Process bone marrow biopsies in as little as 15 to 30 minutes with this rapid hydrochloric acid decalcifier.
  • Process thin pieces of dense bone in 3 to 12 hours
  • Permits utilization of routine end point tests, such as calcium oxalate
  • Protects cells from actions of acids with added ingredients
Documents
Specifications
Specifications
  • Description:
    Epredia™ TBD-1™ decalcifier
  • Size:
    2 x 1 l
Specifications