Order Entry
Canada
Orders LinkContactUsLinkComponent
Wright-Giemsa stain

Wright-Giemsa stain

9.5 L (2.5 gal.)
Catalog # CA84000-186
Supplier:   Thermo Scientific
Supplier #:  89017
Wright-Giemsa stain
Catalog # CA84000-186
Supplier:  Thermo Scientific
CAS Number:  
Wright-Giemsa stain
Wright-Giemsa stain

9.5 L (2.5 gal.)

CAS Number:  
Price
$1,355.95
Log in for availability
Each
Product Details & Documents

Rapidly and conveniently stain blood smears with Epredia™ Wright Giemsa Stain.

Synonyms:  Wright-Giemsa Solution
  • MDL Number:MFCD00132593

Documents

Specifications

Specifications

  • Size:
    9.5 L (2.5 gal.)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.