Order Entry
Canada
Orders LinkContactUsLinkComponent
Hematoxylin modified solution (Harris)

Hematoxylin modified solution (Harris)

473 ml (16 oz.)
Catalog # CA84000-088
Supplier:   Thermo Scientific
Supplier #:  72704
Hematoxylin modified solution (Harris)
Catalog # CA84000-088
Supplier:  Thermo Scientific
CAS Number:  517-28-2
Hematoxylin modified solution (Harris)
Hematoxylin modified solution (Harris)

473 ml (16 oz.)

CAS Number:  517-28-2
Price
$611.65
Log in for availability
Case of 4
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Use Epredia™ Modified Harris Hematoxylin, a regressive nuclear stain, for both histology and cytology.

  • Formulation contains no mercury
  • More stable than conventional Harris Hematoxylin


Epredia™ Modified Harris Hematoxylin is an acidified formulation that provides traditional nuclear staining results. Harris Hematoxylin procedures require acid alcohol differentiation. The nuclear chromatin and cytoplasm are over stained and the acid alcohol is used to destain the cytoplasm and also remove excess staining from the nuclear chromatin. Staining times are typically in the 5 minute range. Our Differentiating Solution can be used successfully with Modified Harris Hematoxylin. Our Epredia Modified Harris Hematoxylin is more stable than most commercial Harris Hematoxylins. It will not abundantly auto-oxidize at the same accelerated rate as other Harris Hematoxylins. The potency of the stain will slowly change, however the staining viability will remain for longer times. The product will crystallize when exposed to colder temperatures. Simply filter the crystals or put the contents of the bottle and crystals on a heated stir plate (35° C) and the crystals will go back into solution. DO NOT BOIL! It is highly recommended to filter Modified Harris Hematoxylin on a daily basis.

Synonyms:  DcH-48, Natural Black I, Hydroxybrasilin
  • CAS Number:517-28-2
  • Formula:C₁₆H₁₄O₆
  • MDL Number:MFCD00078111
  • Molecular Weight:302.28 g/mol
Specifications

Specifications

  • Size:
    473 ml (16 oz.)

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.