Order Entry
Canada
Orders LinkContactUsLinkComponent
WHEATON® Screw Thread Vials, DWK Life Sciences

WHEATON® Screw Thread Vials, DWK Life Sciences

Tubular Vials, 5.00 ml, 22×38 mm, Borosilicate glass, Clear, No, 20-400
Catalog # 10070-712
Supplier #:  W219365
WHEATON® Screw Thread Vials, DWK Life Sciences
Catalog # 10070-712
WHEATON® Screw Thread Vials, DWK Life Sciences
WHEATON® Screw Thread Vials, DWK Life Sciences

Tubular Vials, 5.00 ml, 22×38 mm, Borosilicate glass, Clear, No, 20-400

Price
$1,504.50
New Customer Promotional Price
Log in for availability
Case of 480
Product Details & Documents

The tubular design provides excellent clarity and dimensional consistency from vial to vial.

  • Ideal for lyophilization
  • Alternative to serum bottles and vials
  • Heavy walled glass provides impact resistance
  • Screw thread design eliminates need for crimping tools

These screw thread vials fits with I-Loc™ closure or screw cap with thin flange stopper.

Specifications
  • Cap included:
    No
  • Cap Size:
    20-400
  • Capacity:
    5.00 ml
  • Color:
    Clear
  • Description:
    Tubular Vials
  • Material:
    Borosilicate glass
  • Diameter ×H:
    22×38 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.