WHEATON® Screw Thread Vials, DWK Life Sciences
Tubular Vials, 20.00 ml, 30×52 mm, Borosilicate glass, Amber, No, 20-400
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: W216377
WHEATON® Screw Thread Vials, DWK Life Sciences
Catalog # 10068-206
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
Product Details & Documents
The tubular design provides excellent clarity and dimensional consistency from vial to vial.
- Ideal for lyophilization
- Alternative to serum bottles and vials
- Heavy walled glass provides impact resistance
- Screw thread design eliminates need for crimping tools
These screw thread vials fits with I-Loc™ closure or screw cap with thin flange stopper.
Documents
Certificates
Search by Lot Number10068-210_0000039015 10070-712_0000046520 10070-712_0000063884.pdf 10070-712_1805000061 10070-714_0000032070
More
Wheaton Vial GUide
Product Literature
Specifications
- Cap included:No
- Cap Size:20-400
- Capacity:20.00 ml
- Color:Amber
- Description:Tubular Vials
- Material:Borosilicate glass
- Diameter ×H:30×52 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



